missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M20355G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20355G Supplier TCI America Supplier No. M20355G

Specifications

Chemical Name or Material 3,4-(Methylenedioxy)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 94839-07-3
Melting Point 229°C
Color White-Yellow
Molecular Formula C7H7BO4
Quantity 5 g
MDL Number MFCD01009695
Synonym 3,4-methylenedioxyphenylboronic acid, benzo d 1,3 dioxol-5-ylboronic acid, 3,4-methylenedioxy phenylboronic acid, 3,4-methylenedioxyphenyl boronic acid, 3,4-methylenedioxy benzeneboronic acid, 1,3-benzodioxole-5-boronic acid, 3,4-methylenedioxybenzeneboronic acid, 2h-1,3-benzodioxol-5-yl boronic acid, 2h-1,3-benzodioxol-5-yl-boranediol, 2h-1,3-benzodioxol-5-ylboronic acid
InChI Key CMHPUBKZZPSUIQ-UHFFFAOYSA-N
SMILES B(C1=CC2=C(C=C1)OCO2)(O)O
IUPAC Name 1,3-benzodioxol-5-ylboronic acid
Molecular Weight (g/mol) 165.939
PubChem CID 2734371
Formula Weight 165.94
Physical Form Crystalline Powder
Show More Show Less