missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)phenylacetic acid, 98%

Catalog No. AAA1122906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1122906 5 g
AAA1122914 25 g
AAA1122922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1122906 Supplier Thermo Scientific Chemicals Supplier No. A1122906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2861-28-1 | C9H8O4 | 180.159 g/mol

3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.

Solubility
Soluble in water (partly), ethanol, and methanol.

Notes
Store at room temperature. Store away from oxidizing agents. Store in a cool, dry place. Store in a tightly closed container.

Chemical Identifiers

CAS 2861-28-1
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00014576
InChI Key ODVLMCWNGKLROU-UHFFFAOYSA-N
Synonym 1,3-benzodioxole-5-acetic acid, 3,4-methylenedioxy phenylacetic acid, 2-benzo d 1,3 dioxol-5-yl acetic acid, 3,4-methylenedioxyphenylacetic acid, homopiperonylic acid, benzo-1,3-dioxole-5-acetic acid, 1,3-benzodioxol-5-yl acetic acid, 2-2h-1,3-benzodioxol-5-yl acetic acid, 2-1,3-benzodioxol-5-yl acetic acid, 1,3-benzodioxol-5-ylacetic acid
PubChem CID 76115
IUPAC Name 2-(1,3-benzodioxol-5-yl)acetic acid
SMILES C1OC2=C(O1)C=C(C=C2)CC(=O)O

Specifications

Melting Point 127°C to 130°C
Quantity 5 g
Beilstein 177739
Solubility Information Soluble in water (partly),ethanol,and methanol.
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 3,4- (Methylenedioxy)phenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-679-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only