missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)benzylideneacetone, 98%

Catalog No. AAL0867518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0867518 50 g
AAL0867509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0867518 Supplier Thermo Scientific Chemicals Supplier No. L0867518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3160-37-0 | C11H10O3 | 190.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3160-37-0
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00016907
InChI Key XIYPXOFSURQTTJ-NSCUHMNNSA-N
Synonym piperonalacetone, heliotropyl acetone, acetone, piperonylidene, piperonylideneacetone, piperonylidene acetone, unii-t43byj64eg, 3-buten-2-one, 4-1,3-benzodioxol-5-yl, ccris 6270, 3,4-methylenedioxy benzalacetone, 3,4-methylenedioxy benzylideneacetone
PubChem CID 6040503
IUPAC Name (E)-4-(1,3-benzodioxol-5-yl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC2=C(C=C1)OCO2

Specifications

Melting Point 108°C to 110°C
Quantity 50 g
Beilstein 83983
Formula Weight 190.2
Percent Purity 98%
Chemical Name or Material 3,4-(Methylenedioxy)benzylideneacetone
EINECSNumber 221-608-4
RTECSNumber EM9822500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only