missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)benzeneboronic acid, 98%

Catalog No. p-7027455
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2421703 1 g
AAB2421706 5 g
AAB2421714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2421703 Supplier Thermo Scientific Chemicals Supplier No. B2421703

CAS: 94839-07-3 | C7H7BO4 | 165.939 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94839-07-3
Molecular Formula C7H7BO4
Molecular Weight (g/mol) 165.939
MDL Number MFCD01009695
InChI Key CMHPUBKZZPSUIQ-UHFFFAOYSA-N
Synonym 3,4-methylenedioxyphenylboronic acid, benzo d 1,3 dioxol-5-ylboronic acid, 3,4-methylenedioxy phenylboronic acid, 3,4-methylenedioxyphenyl boronic acid, 3,4-methylenedioxy benzeneboronic acid, 1,3-benzodioxole-5-boronic acid, 3,4-methylenedioxybenzeneboronic acid, 2h-1,3-benzodioxol-5-yl boronic acid, 2h-1,3-benzodioxol-5-yl-boranediol, 2h-1,3-benzodioxol-5-ylboronic acid
PubChem CID 2734371
IUPAC Name 1,3-benzodioxol-5-ylboronic acid
SMILES B(C1=CC2=C(C=C1)OCO2)(O)O

Specifications

Melting Point 239°C to 243°C
Quantity 1 g
Beilstein 5523347
Formula Weight 165.94
Percent Purity 98%
Chemical Name or Material 3,4-(Methylenedioxy)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only