missing translation for 'onlineSavingsMsg'
Learn More

3',4'-(Methylenedioxy)acetophenone, 98%

Catalog No. p-7021315
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1359709 10 g
AAA1359718 50 g
AAA1359730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1359709 Supplier Thermo Scientific Chemicals Supplier No. A1359709

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3162-29-6 | C9H8O3 | 164.16 g/mol

3?,4?-(Methylenedioxy)acetophenone is used in the synthesis of N-ethyl-1-(3,4-methylenedioxyphenyl)ethylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′-(Methylenedioxy)acetophenone is used in the synthesis of N-ethyl-1-(3,4-methylenedioxyphenyl)ethylamine.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 3162-29-6
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00005831
InChI Key BMHMKWXYXFBWMI-UHFFFAOYSA-N
Synonym 3',4'-methylenedioxy acetophenone, 1-benzo d 1,3 dioxol-5-yl ethanone, 5-acetyl-1,3-benzodioxole, 3,4-methylenedioxyacetophenone, 3,4-methylenedioxy acetophenone, 3',4'-methylenedioxyacetophenone, 1-1,3-benzodioxol-5-yl ethanone, acetopiperone, 1-1,3-benzodioxol-5-yl ethan-1-one, ethanone, 1-1,3-benzodioxol-5-yl
PubChem CID 76622
IUPAC Name 1-(1,3-benzodioxol-5-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)OCO2

Specifications

Melting Point 85°C to 89°C
Boiling Point 152°C to 155°C (15 mmHg)
Quantity 10 g
Beilstein 139669
Solubility Information Slightly soluble in water.
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material 3',4'-(Methylenedioxy)acetophenone
EINECSNumber 221-613-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only