missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 97%

Catalog No. AAH2922406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2922406 5 g
AAH2922403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2922406 Supplier Thermo Scientific Chemicals Supplier No. H2922406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 883738-38-3 | C18H27BN2O3 | 330.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 883738-38-3
Molecular Formula C18H27BN2O3
Molecular Weight (g/mol) 330.24
MDL Number MFCD05864308
InChI Key ZYDMNNQZEUSDGG-UHFFFAOYSA-N
Synonym 4-methylpiperazin-1-yl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 3-4-methyl-1-piperazinylcarbonyl benzeneboronic acid pinacol ester, 3-4-methylpiperazine-1-carbonyl phenylboronic acid, pinacol ester, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzamide, 3-4-methylpiperazine-1-carbonyl phenyl boronic acid pinacol ester, 1-methyl-4-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, 1-methyl-4-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, n-4-methylpiperazine-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb046
PubChem CID 2760029
SMILES CN1CCN(CC1)C(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 86°C to 89°C
Quantity 5 g
IUPAC Name 1-methyl-4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine
Formula Weight 330.23
Percent Purity 97%
Chemical Name or Material 3-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only