missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Methyl-1-piperazinyl)aniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 85°C |
| Quantity | 1 g |
| UN Number | 3263 |
| Formula Weight | 191.28 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-(4-Methyl-1-piperazinyl)aniline |
Safety and Handling
| TSCA | No |