missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester, 97%

Catalog No. p-7030418
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
AAH28557MD 250 mg
AAH2855703 1 g
2 options

Catalog No. AAH28557MD

Supplier: Thermo Scientific Chemicals H28557MD

Only null left

CAS: 1073371-72-8 | C16H25BO3 | 276.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073371-72-8
Molecular Formula C16H25BO3
Molecular Weight (g/mol) 276.183
MDL Number MFCD09953503
InChI Key ZJNJYHQBUIHZLN-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-1-propylboronic acid pinacol ester, 2-3-4-methoxyphenyl propyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4-methoxyphenyl propylboronic acid pinacol ester, 3-4-methoxyphenyl propylboronic acid,pinacol ester, 3-4-methoxyphenyl propylboronic acid, pinacol ester
PubChem CID 46739066
IUPAC Name 2-[3-(4-methoxyphenyl)propyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCCC2=CC=C(C=C2)OC

Specifications

Quantity 250 mg
Formula Weight 276.19
Percent Purity 97%
Chemical Name or Material 3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only