missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxybenzoyl)propionic Acid 98.0+%, TCI America™

Catalog No. M13835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13835G 5 g
M138325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13835G Supplier TCI America Supplier No. M13835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3153-44-4
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00002795
InChI Key OMTDIBZSUZNVJK-UHFFFAOYSA-N
Synonym 3-4-methoxybenzoyl propionic acid, 4-4-methoxyphenyl-4-oxobutanoic acid, 4-4-methoxyphenyl-4-oxobutyric acid, 3-4-methoxybenzoyl propanoic acid, beta-p-methoxybenzoyl propionic acid, 4-4-methoxyphenyl-4-oxo-butyric acid, 4-4-methoxyphenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-methoxy-gamma-oxo, maybridge1_002003, acmc-1cl1u
PubChem CID 76613
IUPAC Name 4-(4-methoxyphenyl)-4-oxobutanoic acid
SMILES COC1=CC=C(C=C1)C(=O)CCC(=O)O

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 5 g
Formula Weight 208.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Methoxybenzoyl)propionic Acid