missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxybenzoyl)propionic acid, 98+%

Catalog No. AAA1821014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1821014 25 g
AAA1821006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1821014 Supplier Thermo Scientific Chemicals Supplier No. A1821014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 3153-44-4 | C11H12O4 | 208.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3153-44-4
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00002795
InChI Key OMTDIBZSUZNVJK-UHFFFAOYSA-N
Synonym 3-4-methoxybenzoyl propionic acid, 4-4-methoxyphenyl-4-oxobutanoic acid, 4-4-methoxyphenyl-4-oxobutyric acid, 3-4-methoxybenzoyl propanoic acid, beta-p-methoxybenzoyl propionic acid, 4-4-methoxyphenyl-4-oxo-butyric acid, 4-4-methoxyphenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-methoxy-gamma-oxo, maybridge1_002003, acmc-1cl1u
PubChem CID 76613
IUPAC Name 4-(4-methoxyphenyl)-4-oxobutanoic acid
SMILES COC1=CC=C(C=C1)C(=O)CCC(=O)O

Specifications

Melting Point 147°C to 151°C
Quantity 25 g
Beilstein 785074
Formula Weight 208.21
Percent Purity ≥98%
Chemical Name or Material 3-(4-Methoxybenzoyl)propionic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 221-593-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only