missing translation for 'onlineSavingsMsg'
Learn More

3,4-Lutidine 98.0+%, TCI America™

Catalog No. L0066500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0066500ML 500 mL
L006625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0066500ML Supplier TCI America Supplier No. L0066500ML

Chemical Identifiers

CAS 583-58-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00006403
InChI Key NURQLCJSMXZBPC-UHFFFAOYSA-N
PubChem CID 11417
IUPAC Name 3,4-dimethylpyridine
SMILES CC1=C(C=NC=C1)C

Specifications

Melting Point -6°C
Color Yellow
Boiling Point 179°C
Quantity 500 mL
UN Number 2810
Synonym 3,4-lutidine, pyridine, 3,4-dimethyl, unii-fcu895kj7h, pyridine,3,4-dimethyl, fcu895kj7h, 3,4-lutidene, t 3,4-lutidine, 3,4-dimethyl-pyridine, 5-20-06-00054 beilstein handbook reference
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Lutidine
EINECSNumber (5)-0712
RTECSNumber OK9800000
TSCA Yes