missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 3/4 in. ID Equal Tee - Natural Kynar - QC

Catalog No. 01000647
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01000647 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01000647

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing.

  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661
  • USP81
  • USP 8
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Complaince Certificates Available
Description Class 7 Cleanroom Manufactured
Material Kynar PVDF
For Use With (Application) Tube to Tube Connector for Medical, Pharmaceutical, Industrial applications
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Natural
Product Type Equal Tee
Quantity 100 Pk.