Learn More
3-(4-Hydroxyphenyl)propionic acid, 99%
CAS: 501-97-3 | C9H10O3 | 166.176 g/mol
Supplier: Thermo Scientific Chemicals A1456718
Description
3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 3-(4-Hydroxyphenyl)propionic acid | |
| 126°C to 131°C | |
| MFCD00002778 | |
| 2209841 | |
| Soluble in water. | |
| C1=CC(=CC=C1CCC(=O)O)O | |
| 166.176 | |
| CHEBI:32980 | |
| 99% |
| 501-97-3 | |
| C9H10O3 | |
| 50 g | |
| 3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid | |
| NMHMNPHRMNGLLB-UHFFFAOYSA-N | |
| 3-(4-hydroxyphenyl)propanoic acid | |
| 10394 | |
| 166.18 |
Chemical Identifiers
| 501-97-3 | |
| 166.176 | |
| NMHMNPHRMNGLLB-UHFFFAOYSA-N | |
| 10394 | |
| 3-(4-hydroxyphenyl)propanoic acid |
| C9H10O3 | |
| MFCD00002778 | |
| 3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid | |
| CHEBI:32980 | |
| C1=CC(=CC=C1CCC(=O)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-931-3
RTECSNumber : MW5342000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only