Learn More
3,4-Ethylenedioxythiophene, 99%
CAS: 126213-50-1 | C6H6O2S | 142.18 g/mol
Supplier: Thermo Scientific Chemicals 454930250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3,4-Ethylenedioxythiophene | |
| 10.5°C to 12.0°C | |
| 193.0°C | |
| Authentic | |
| C6H6O2S | |
| 1.331 | |
| GKWLILHTTGWKLQ-UHFFFAOYSA-N | |
| 2,3-dihydrothieno[3,4-b][1,4]dioxine | |
| 4421864 | |
| 99% |
| 126213-50-1 | |
| 1.3310g/mL | |
| 109°C | |
| 98.5% min. (GC) | |
| 25 g | |
| 3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane | |
| C1COC2=CSC=C2O1 | |
| 142.18 | |
| 142.18 |
Chemical Identifiers
| 126213-50-1 | |
| 142.18 | |
| 3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane | |
| 2,3-dihydrothieno[3,4-b][1,4]dioxine |
| C6H6O2S | |
| GKWLILHTTGWKLQ-UHFFFAOYSA-N | |
| 4421864 | |
| C1COC2=CSC=C2O1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
GHS Signal Word: Danger
RUO – Research Use Only