missing translation for 'onlineSavingsMsg'
Learn More

3,4-Ethylenedioxythiophene, 97%

Catalog No. p-7037730
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5653306 5 g
AAH5653314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5653306 Supplier Thermo Scientific Chemicals Supplier No. H5653306
Only null left

CAS: 126213-50-1 | C6H6O2S | 142.172 g/mol

3,4-Ethylenedioxythiophene is used as a monomer to synthesize the conductive polymers and used as a reductant in the one-pot synthesis of gold nanoparticles from chloroauric acid, as starting material used in palladium-catalyzed mono and bis-arylation reactions and in the synthesis of conjugated polymers and copolymers, with potential optical applications. It is also used in redox activity, electroactivity and conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Ethylenedioxythiophene is used as a monomer to synthesize the conductive polymers and used as a reductant in the one-pot synthesis of gold nanoparticles from chloroauric acid, as starting material used in palladium-catalyzed mono and bis-arylation reactions and in the synthesis of conjugated polymers and copolymers, with potential optical applications. It is also used in redox activity, electroactivity and conductivity.

Solubility
Immsicible with water. Miscible with alcohol and ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 126213-50-1
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.172
MDL Number MFCD02093622
InChI Key GKWLILHTTGWKLQ-UHFFFAOYSA-N
Synonym 3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane
PubChem CID 4421864
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxine
SMILES C1COC2=CSC=C2O1

Specifications

Density 1.331
Boiling Point 193°C
Flash Point 104°C (219°F)
Refractive Index 1.5765
Quantity 5 g
UN Number UN2810
Solubility Information Immsicible with water. Miscible with alcohol and ether.
Formula Weight 142.17
Percent Purity 97%
Chemical Name or Material 3,4-Ethylenedioxythiophene
Hazard Category H302-H311-H319
Hazard Statement GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 603-128-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only