missing translation for 'onlineSavingsMsg'
Learn More

3,4-Ethylenedioxyaniline 98.0+%, TCI America™

Catalog No. E038525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E038525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E038525ML Supplier TCI America Supplier No. E038525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22013-33-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00006824
InChI Key BZKOZYWGZKRTIB-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-amine, 3,4-ethylenedioxyaniline, 6-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-6-amine, 6-amino-1,4-benzodioxan, 1,4-benzodioxan-6-ylamine, 1,4-benzodioxin-6-amine, 2,3-dihydro, 3,4-ethylenedioxy aniline, 2,3-dihydro-benzo 1,4 dioxin-6-ylamine, 2,3-dihydro-1,4-benzodioxin-6-amin
PubChem CID 89148
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-amine
SMILES NC1=CC=C2OCCOC2=C1

Specifications

Boiling Point 162°C
Quantity 25 mL
Formula Weight 151.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3,4-Ethylenedioxyaniline