missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylcyclohexanol (mixture of isomers) 97.0+%, TCI America™

Catalog No. D070325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D070325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D070325ML Supplier TCI America Supplier No. D070325ML

Chemical Identifiers

CAS 5715-23-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001506
InChI Key ZBAXJUPCYVIBSP-UHFFFAOYSA-N
PubChem CID 97960
IUPAC Name 3,4-dimethylcyclohexan-1-ol
SMILES CC1CCC(CC1C)O

Specifications

Color Colorless
Boiling Point 189°C
Quantity 25 mL
Formula Weight 128.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethylcyclohexanol (mixture of isomers)