missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-2-hexanol, erythro + threo, 98%

Catalog No. AAB2024303
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2024303 1 g
AAB2024314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2024303 Supplier Thermo Scientific Chemicals Supplier No. B2024303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19550-05-1 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19550-05-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00021887
InChI Key LBJWJHMCZHKLQU-UHFFFAOYSA-N
Synonym 3,4-dimethyl-2-hexanol, 2-hexanol, 3,4-dimethyl, 3,4-dimethyl-2-hexanol, erythro + threo, 3,4-dimethyl-2-hexanol mixture of isomers, 2-hexanol,4-dimethyl, acmc-1ah66, 3,4-dimethyl-2-hexanol mixture of isomers a, 3,4-dimethyl-2-hexanol, threo + erythro
PubChem CID 140547
IUPAC Name 3,4-dimethylhexan-2-ol
SMILES CCC(C)C(C)C(C)O

Specifications

Quantity 1 g
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 3,4-Dimethyl-2-hexanol, erythro + threo
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 631-563-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only