missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-1-pentyn-3-ol 96.0+%, TCI America™

Catalog No. D127610ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D127610ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D127610ML Supplier TCI America Supplier No. D127610ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1482-15-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00039845
InChI Key DZNLEQBXXLGELU-UHFFFAOYNA-N
Synonym 3,4-dimethyl-1-pentyn-3-ol, 1-pentyn-3-ol, 3,4-dimethyl, acmc-1bzc3, 2-isopropyl-3-butyne-2-ol, 1-pentyn-3-ol,4-dimethyl, 3,4-dimethyl-pent-1-yn-3-ol, dznleqbxxlgelu-uhfffaoysa, 1-pentyn-3-ol,3,4-dimethyl, 1,2-dimethyl-1-hydroxypropyl acetylene
PubChem CID 95483
IUPAC Name 3,4-dimethylpent-1-yn-3-ol
SMILES CC(C)C(C)(O)C#C

Specifications

Color Yellow
Boiling Point 133°C
Quantity 10 mL
UN Number 1987
Formula Weight 112.17
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethyl-1-pentyn-3-ol