missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-1-pentyn-3-ol, 94%

Catalog No. AAA1990606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1990606 5 g
AAA1990614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1990606 Supplier Thermo Scientific Chemicals Supplier No. A1990606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1482-15-1 | C7H12O | 112.17 g/mol

3,4-Dimethyl-1-pentyn-3-ol is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethyl-1-pentyn-3-ol is used as pharmaceutical intermediates.

Solubility
Fully miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.

Chemical Identifiers

CAS 1482-15-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00039845
InChI Key DZNLEQBXXLGELU-UHFFFAOYNA-N
PubChem CID 95483
IUPAC Name 3,4-dimethylpent-1-yn-3-ol
SMILES CC(C)C(C)(O)C#C

Specifications

Density 0.87
Boiling Point 133°C
Flash Point 40°C (104°F)
Quantity 5 g
UN Number UN1987
Synonym 3,4-dimethyl-1-pentyn-3-ol, 1-pentyn-3-ol, 3,4-dimethyl, acmc-1bzc3, 2-isopropyl-3-butyne-2-ol, 1-pentyn-3-ol,4-dimethyl, 3,4-dimethyl-pent-1-yn-3-ol, dznleqbxxlgelu-uhfffaoysa, 1-pentyn-3-ol,3,4-dimethyl, 1,2-dimethyl-1-hydroxypropyl acetylene
Solubility Information Fully miscible in water.
Formula Weight 112.17
Percent Purity 94%
Chemical Name or Material 3,4-Dimethyl-1-pentyn-3-ol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only