missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxythiophene 98.0+%, TCI America™

Catalog No. D27281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27281G 1 g
D27285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27281G Supplier TCI America Supplier No. D27281G

Chemical Identifiers

CAS 51792-34-8
Molecular Formula C6H8O2S
Molecular Weight (g/mol) 144.188
MDL Number MFCD01096546
InChI Key ZUDCKLVMBAXBIF-UHFFFAOYSA-N
Synonym 3,4-dimethoxy thiophene, thiophene, 3,4-dimethoxy, pubchem15752, 3,4-dimethoxy-thiophene, acmc-209kw1, ksc491i4n, 3,4-dimethoxythiophene
PubChem CID 3613501
IUPAC Name 3,4-dimethoxythiophene
SMILES COC1=CSC=C1OC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 144.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethoxythiophene