missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7193873
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35125G Supplier TCI America Supplier No. D35125G
Only null left

Chemical Identifiers

CAS 122775-35-3
Molecular Formula C8H11BO4
MDL Number MFCD01074574
InChI Key RCVDPBFUMYUKPB-UHFFFAOYSA-N
PubChem CID 2734702
IUPAC Name (3,4-dimethoxyphenyl)boronic acid

Specifications

Melting Point 250°C
Quantity 5 g
Synonym 3,4-dimethoxyphenyl boronic acid, 3,4-dimethoxybenzeneboronic acid, 3,4-dimethoxyphenyl boranediol, 3,4-dimethoxybenzene boronic acid, boronic acid, 3,4-dimethoxyphenyl, 3,4-dimethoxyphenylboronicacid, 3, 4-dimethoxyphenylboronic acid, pubchem1826, acmc-209amm, ksc174q5t
SMILES COC1=CC=C(C=C1OC)B(O)O
Molecular Weight (g/mol) 181.98
Formula Weight 181.98
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride)