missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyflavone 98.0+%, TCI America™

Catalog No. D4794200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4794200MG 200 mg
D47941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4794200MG Supplier TCI America Supplier No. D4794200MG
Only null left

Chemical Identifiers

CAS 4143-62-8
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00143009
InChI Key ZGHORMOOTZTQFL-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyflavone, 2-3,4-dimethoxyphenyl-4h-chromen-4-one, 2-3,4-dimethoxyphenyl chromen-4-one, chembl91153, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl, 3',4'-dmf, pubchem9847, spectrum_001039, acmc-1apwv, specplus_000555
PubChem CID 688674
IUPAC Name 2-(3,4-dimethoxyphenyl)chromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2)OC

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 200 mg
Formula Weight 282.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dimethoxyflavone
RTECSNumber DJ3012950
TSCA No