missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer

CAS: 2316-26-9 | C11H12O4 | 208.21 g/mol

$50.71 - $118.54

Chemical Identifiers

CAS 2316-26-9
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.21
MDL Number MFCD00004387
InChI Key HJBWJAPEBGSQPR-GQCTYLIASA-N
Synonym 3,4-dimethoxycinnamic acid, dimethylcaffeic acid, caffeic acid dimethyl ether, 2e-3-3,4-dimethoxyphenyl prop-2-enoic acid, dimethyl caffeic acid, e-3-3,4-dimethoxyphenyl acrylic acid, e-3,4-dimethoxycinnamic acid, unii-bvz841pvjl, 2e-3-3,4-dimethoxyphenyl acrylic acid, 3-3,4-dimethoxyphenyl acrylic acid
PubChem CID 717531
ChEBI CHEBI:86549
IUPAC Name (E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=C(C=C1)C=CC(=O)O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC115510050
SDS View Documents
Thermo Scientific Chemicals
115510050
5 g Glass bottle
Each for $50.71
Only null left
 
AC115510250
SDS View Documents
Thermo Scientific Chemicals
115510250
25 g Glass bottle
Each for $118.54
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2316-26-9
208.21
HJBWJAPEBGSQPR-GQCTYLIASA-N
717531
(E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
C11H12O4
MFCD00004387
3,4-dimethoxycinnamic acid, dimethylcaffeic acid, caffeic acid dimethyl ether, 2e-3-3,4-dimethoxyphenyl prop-2-enoic acid, dimethyl caffeic acid, e-3-3,4-dimethoxyphenyl acrylic acid, e-3,4-dimethoxycinnamic acid, unii-bvz841pvjl, 2e-3-3,4-dimethoxyphenyl acrylic acid, 3-3,4-dimethoxyphenyl acrylic acid
CHEBI:86549
COC1=C(C=C(C=C1)C=CC(=O)O)OC

Specifications

2316-26-9
100.0
Yellow to Beige
98.5% min. (GC)
C11H12O4
MFCD00004387
10, 437
HJBWJAPEBGSQPR-GQCTYLIASA-N
(E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
717531
208.21
Powder
98.5
181°C to 184°C
Authentic
Glass bottle
(CH3O)2C6H3CH=CHCO2H
5 g
3,4-dimethoxycinnamic acid, dimethylcaffeic acid, caffeic acid dimethyl ether, 2e-3-3,4-dimethoxyphenyl prop-2-enoic acid, dimethyl caffeic acid, e-3-3,4-dimethoxyphenyl acrylic acid, e-3,4-dimethoxycinnamic acid, unii-bvz841pvjl, 2e-3-3,4-dimethoxyphenyl acrylic acid, 3-3,4-dimethoxyphenyl acrylic acid
COC1=C(C=C(C=C1)C=CC(=O)O)OC
208.21
CHEBI:86549
99%
3, 4-Dimethoxycinnamic acid, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 219-025-5

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only