missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzeneboronic acid, 98%

Catalog No. AAB2424014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2424014 25 g
AAB2424003 1 g
AAB2424006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2424014 Supplier Thermo Scientific Chemicals Supplier No. B2424014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 122775-35-3 | C8H11BO4 | 181.98 g/mol

3,4-Dimethoxybenzeneboronic acid is used in organic light-emitting diode. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxybenzeneboronic acid is used in organic light-emitting diode. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (25 g/L).

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 122775-35-3
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.98
MDL Number MFCD01074574
InChI Key RCVDPBFUMYUKPB-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenyl boronic acid, 3,4-dimethoxybenzeneboronic acid, 3,4-dimethoxyphenyl boranediol, 3,4-dimethoxybenzene boronic acid, boronic acid, 3,4-dimethoxyphenyl, 3,4-dimethoxyphenylboronicacid, 3, 4-dimethoxyphenylboronic acid, pubchem1826, acmc-209amm, ksc174q5t
PubChem CID 2734702
IUPAC Name (3,4-dimethoxyphenyl)boronic acid
SMILES COC1=CC=C(C=C1OC)B(O)O

Specifications

Melting Point ∼240°C
Quantity 25 g
Beilstein 5334877
Solubility Information Soluble in water (25g/L).
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only