missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxybenzaldehyde 98.0+%, TCI America™
Supplier: TCI America D0507500G
Specifications
| 3,4-Dimethoxybenzaldehyde | |
| 43°C | |
| C9H10O3 | |
| 500 g | |
| WJUFSDZVCOTFON-UHFFFAOYSA-N | |
| 3,4-dimethoxybenzaldehyde | |
| 8419 | |
| 166.18 | |
| Crystalline Powder |
| 120-14-9 | |
| 281°C | |
| MFCD00003363 | |
| veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin | |
| COC1=CC=C(C=O)C=C1OC | |
| 166.18 | |
| CHEBI:17098 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 120-14-9 | |
| 166.18 | |
| WJUFSDZVCOTFON-UHFFFAOYSA-N | |
| 8419 | |
| 3,4-dimethoxybenzaldehyde |
| C9H10O3 | |
| MFCD00003363 | |
| veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin | |
| CHEBI:17098 | |
| COC1=CC=C(C=O)C=C1OC |
Safety and Handling
EINECSNumber : (3)-1203&(3)-2665
RTECSNumber : YX5088000
TSCA : Yes