missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. D050725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D050725G 25 g
D0507100G 100 g
D0507500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D050725G Supplier TCI America Supplier No. D050725G

Chemical Identifiers

CAS 120-14-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00003363
InChI Key WJUFSDZVCOTFON-UHFFFAOYSA-N
Synonym veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin
PubChem CID 8419
ChEBI CHEBI:17098
IUPAC Name 3,4-dimethoxybenzaldehyde
SMILES COC1=CC=C(C=O)C=C1OC

Specifications

Melting Point 43°C
Boiling Point 281°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxybenzaldehyde
EINECSNumber (3)-1203&(3)-2665
RTECSNumber YX5088000
TSCA Yes