missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzaldehyde, 99+%

CAS: 120-14-9 | C9H10O3 | 166.18 g/mol

Supplier:  Thermo Scientific Chemicals 162611000

 View more versions of this product

Catalog No. AC162611000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3, 4-Dimethoxybenzaldehyde
120-14-9
100.0
White to Yellow
>112°C
99% min. (GC)
C9H10O3
MFCD00003363
08, 255
veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin
WJUFSDZVCOTFON-UHFFFAOYSA-N
3,4-dimethoxybenzaldehyde
8419
166.18
Lumps or Fused Solid
99+%
99.0
42°C to 45°C
281°C
Authentic
Plastic bottle
(CH3O)2C6H3CHO
100 g
15, 10145
Solubility in water: soluble in hot water. Other solubilities: freely soluble in alcohol and ether
COC1=CC=C(C=O)C=C1OC
166.18
CHEBI:17098
99+%

Chemical Identifiers

120-14-9
166.18
WJUFSDZVCOTFON-UHFFFAOYSA-N
8419
3,4-dimethoxybenzaldehyde
C9H10O3
MFCD00003363
veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin
CHEBI:17098
COC1=CC=C(C=O)C=C1OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-373-2

RTECSNumber : YX5088000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only