missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyacetophenone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 168851000

 View more versions of this product

Catalog No. AC168851000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3', 4'-Dimethoxyacetophenone
97.5
286.0°C to 288.0°C
Authentic
Glass bottle
(CH3O)2C6H3COCH3
100g
3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
IQZLUWLMQNGTIW-UHFFFAOYSA-N
1-(3,4-dimethoxyphenyl)ethanone
14328
180.2
1131-62-0
100.0
>112°C
97.5% min. (GC)
C10H12O3
MFCD00008737
08, II, 298
Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol
CC(=O)C1=CC(=C(C=C1)OC)OC
180.2
CHEBI:86576
98%

Chemical Identifiers

1131-62-0
180.2
IQZLUWLMQNGTIW-UHFFFAOYSA-N
14328
1-(3,4-dimethoxyphenyl)ethanone
C10H12O3
MFCD00008737
3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
CHEBI:86576
CC(=O)C1=CC(=C(C=C1)OC)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautious

GHS Signal Word: Warning

EINECSNumber : 214-468-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only