missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyacetophenone, 98+%

CAS: 1131-62-0 | C10H12O3 | 180.203 g/mol

Supplier:  Thermo Scientific Chemicals L0273730

 View more versions of this product

Catalog No. AAL0273730


Only null left
Explore available promotions

Description

Description

3',4'-Dimethoxyacetophenone is used as Catalytic agent; Petrochemical additive. It is also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′-Dimethoxyacetophenone is used as Catalytic agent; Petrochemical additive. It is also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Protect from moisture, heat. Incompatible with Strong oxidizing agents. Protect from direct light.
Specifications

Specifications

3',4'-Dimethoxyacetophenone
47°C to 51°C
>110°C (230°F)
C10H12O3
250 g
3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
IQZLUWLMQNGTIW-UHFFFAOYSA-N
1-(3,4-dimethoxyphenyl)ethanone
14328
180.2
1131-62-0
286°C to 288°C
Odorless
MFCD00008737
781213
Soluble in methanol. Insoluble in water.
CC(=O)C1=CC(=C(C=C1)OC)OC
180.203
CHEBI:86576
≥98%

Chemical Identifiers

1131-62-0
180.203
IQZLUWLMQNGTIW-UHFFFAOYSA-N
14328
1-(3,4-dimethoxyphenyl)ethanone
C10H12O3
MFCD00008737
3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
CHEBI:86576
CC(=O)C1=CC(=C(C=C1)OC)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

EINECSNumber : 214-468-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only