missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyacetophenone, 98+%

Catalog No. AAL0273718
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0273718 50 g
AAL0273730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0273718 Supplier Thermo Scientific Chemicals Supplier No. L0273718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1131-62-0 | C10H12O3 | 180.203 g/mol

3',4'-Dimethoxyacetophenone is used as Catalytic agent; Petrochemical additive. It is also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′-Dimethoxyacetophenone is used as Catalytic agent; Petrochemical additive. It is also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Protect from moisture, heat. Incompatible with Strong oxidizing agents. Protect from direct light.

Chemical Identifiers

CAS 1131-62-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00008737
InChI Key IQZLUWLMQNGTIW-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
PubChem CID 14328
ChEBI CHEBI:86576
IUPAC Name 1-(3,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)OC

Specifications

Melting Point 47°C to 51°C
Boiling Point 286°C to 288°C
Flash Point >110°C (230°F)
Odor Odorless
Quantity 50 g
Beilstein 781213
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material 3',4'-Dimethoxyacetophenone
Hazard Category H302-H319
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 214-468-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only