missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dihydroxypropiophenone, 98%

Catalog No. p-7021205
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1344403 1 g
AAA1344406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1344403 Supplier Thermo Scientific Chemicals Supplier No. A1344403

CAS: 7451-98-1 | C9H10O3 | 166.176 g/mol

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 7451-98-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016440
InChI Key HNWIHBDMOYWCGX-UHFFFAOYSA-N
Synonym 3',4'-dihydroxypropiophenone, 3,4-dihydroxypropiophenone, propiophenone, 3',4'-dihydroxy, 1-3,4-dihydroxyphenyl propan-1-one, 1-propanone, 1-3,4-dihydroxyphenyl, 1-3,4-dihydroxyphenyl-1-propanone, 1-propanone,1-3,4-dihydroxyphenyl, 4-propionylbrenzcatechin, propiophenone,4'-dihydroxy, 3', 4'-dihydroxypropiophenone
PubChem CID 96267
IUPAC Name 1-(3,4-dihydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC(=C(C=C1)O)O

Specifications

Melting Point 145°C to 146°C
Quantity 1 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 3',4'-Dihydroxypropiophenone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only