missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dihydroxyflavone 97.0+%, TCI America™

Catalog No. D4884200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4884200MG 200 mg
D48841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4884200MG Supplier TCI America Supplier No. D4884200MG
Only null left

Chemical Identifiers

CAS 4143-64-0
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00017600
InChI Key SRNPMQHYWVKBAV-UHFFFAOYSA-N
Synonym 3',4'-dihydroxyflavone, 2-3,4-dihydroxyphenyl-4h-chromen-4-one, 2-3,4-dihydroxyphenyl chromen-4-one, unii-koh101s66v, 3',4'-dihydroxy flavone, 3,4-dihydroxyflavone, 4h-1-benzopyran-4-one, 2-3,4-dihydroxyphenyl, dihydroxyflavone, 3',4', 4hlm, pubchem9842
PubChem CID 145726
IUPAC Name 2-(3,4-dihydroxyphenyl)chromen-4-one
SMILES C1=CC=C2C(=C1)C(=O)C=C(O2)C3=CC(=C(C=C3)O)O

Specifications

Melting Point 248°C
Quantity 200 mg
Formula Weight 254.24
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dihydroxyflavone