missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxy-3-cyclobutene-1,2-dione 98.0+%, TCI America™

Catalog No. D13995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13995G 5 g
D139925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13995G Supplier TCI America Supplier No. D13995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2892-51-5
Molecular Formula C4H2O4
Molecular Weight (g/mol) 114.056
MDL Number MFCD00001334
InChI Key PWEBUXCTKOWPCW-UHFFFAOYSA-N
Synonym squaric acid, 3,4-dihydroxy-3-cyclobutene-1,2-dione, quadratic acid, 3-cyclobutene-1,2-dione, 3,4-dihydroxy, quadratsaeure, cyclobutenedione, dihydroxy, 1,2-diketo-3,4-dihydroxycyclobutene, dihydroxycyclobutenedione, unii-svr9d0vodw, dihydroxycyclobut-3-ene-1,2-dione
PubChem CID 17913
ChEBI CHEBI:52141
IUPAC Name 3,4-dihydroxycyclobut-3-ene-1,2-dione
SMILES C1(=C(C(=O)C1=O)O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 114.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydroxy-3-cyclobutene-1,2-dione
EINECSNumber (3)-4169
RTECSNumber GU1800000
TSCA Yes