missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxy-3-cyclobutene-1,2-dione, 99%

Catalog No. AC154020250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154020250 25 g Glass bottle
AC154020050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154020250 Supplier Thermo Scientific Chemicals Supplier No. 154020250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2892-51-5 | C4H2O4 | 114.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2892-51-5
MDL Number MFCD00001334
InChI Key PWEBUXCTKOWPCW-UHFFFAOYSA-N
Synonym squaric acid, 3,4-dihydroxy-3-cyclobutene-1,2-dione, quadratic acid, 3-cyclobutene-1,2-dione, 3,4-dihydroxy, quadratsaeure, cyclobutenedione, dihydroxy, 1,2-diketo-3,4-dihydroxycyclobutene, dihydroxycyclobutenedione, unii-svr9d0vodw, dihydroxycyclobut-3-ene-1,2-dione
PubChem CID 17913
ChEBI CHEBI:52141
IUPAC Name 3,4-dihydroxycyclobut-3-ene-1,2-dione
SMILES C1(=C(C(=O)C1=O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 293°C
Color White to Beige
Density 190g/mL
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C4H2O4
Quantity 25 g
Beilstein 08, IV, 2701
Fieser 04,466; 13,284; 15,296
Merck Index 15, 8897
Solubility Information Solubility in water: 20g/L (30°C)
Molecular Weight (g/mol) 114.06
Formula Weight 114.06
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dihydroxy-3-cyclobutene-1, 2-dione
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-761-4

RUO – Research Use Only