missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydrocoumarin 98.0+%, TCI America™

Catalog No. D1223500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1223500G 500 g
D122325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1223500G Supplier TCI America Supplier No. D1223500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119-84-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00006881
InChI Key VMUXSMXIQBNMGZ-UHFFFAOYSA-N
Synonym dihydrocoumarin, 3,4-dihydrocoumarin, hydrocoumarin, chroman-2-one, benzodihydropyrone, melilotin, melilotol, 2-chromanone, 1,2-benzodihydropyrone, melilotic lactone
PubChem CID 660
ChEBI CHEBI:16151
IUPAC Name 3,4-dihydrochromen-2-one
SMILES C1CC(=O)OC2=CC=CC=C21

Specifications

Color White
Boiling Point 149°C
Quantity 500 g
Formula Weight 148.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,4-Dihydrocoumarin
EINECSNumber (5)-0689
RTECSNumber MW5775000
TSCA Yes