missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-7-hydroxy-2(1H)-quinolinone 98.0+%, TCI America™

Catalog No. p-7127816
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35995G 5 g
D359925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35995G Supplier TCI America Supplier No. D35995G

Chemical Identifiers

CAS 22246-18-0
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD06410891
InChI Key LKLSFDWYIBUGNT-UHFFFAOYSA-N
Synonym 3,4-dihydro-7-hydroxy-2 1h-quinolinone, 7-hydroxy-3,4-dihydroquinolin-2 1h-one, 7-hydroxy-3,4-dihydro-2 1h-quinolinone, 3,4-dihydro-7-hydroxycarbostyril, 7-hydroxy-3,4-dihydro-quinolin-2-one, 7-hydroxy-3,4-dihydrocarbostyril, 7-hydroxy-3,4-dihydro carbostyril, 7-hydroxy-1,2,3,4-tetrahydroquinolin-2-one, 2 1h-quinolinone, 3,4-dihydro-7-hydroxy, 7-hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline
PubChem CID 2785758
IUPAC Name 7-hydroxy-3,4-dihydro-1H-quinolin-2-one
SMILES C1CC(=O)NC2=C1C=CC(=C2)O

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydro-7-hydroxy-2(1H)-quinolinone