missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-4-oxo-1,2,3-benzotriazine 98.0+%, TCI America™

Catalog No. K00035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00035G 5 g
K000325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00035G Supplier TCI America Supplier No. K00035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-16-4
Molecular Formula C7H5N3O
Molecular Weight (g/mol) 147.137
MDL Number MFCD00052387
InChI Key DMSSTTLDFWKBSX-UHFFFAOYSA-N
Synonym 1,2,3-benzotriazin-4 3h-one, 1,2,3-benzotriazin-4 1h-one, benzazimidone, benzazimide, 1,2,3-benzotriazin-4-ol, 4-ketobenzotriazine, benzoketotriazine, usaf ma-2, 3h-1,2,3-benzotriazin-4-one, 4-ketobenz-1,2,3-triazine
PubChem CID 7006
IUPAC Name 1H-1,2,3-benzotriazin-4-one
SMILES C1=CC=C2C(=C1)C(=O)N=NN2

Specifications

Melting Point 218°C
Quantity 5 g
Formula Weight 147.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydro-4-oxo-1,2,3-benzotriazine
RTECSNumber DM0800000
TSCA Yes