missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyran 97.0+%, TCI America™

Catalog No. D0555100ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0555100ML 100 mL
D055525ML 25 mL
D0555500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0555100ML Supplier TCI America Supplier No. D0555100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-87-2
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00006558
InChI Key BUDQDWGNQVEFAC-UHFFFAOYSA-N
Synonym dihydropyran, 3,4-dihydropyran, 2,3-dihydropyran, 2h-3,4-dihydropyran, 2,3-dihydro-4h-pyran, dihydro-2h-pyran, 2h-pyran, dihydro, 2h-pyran, 3,4-dihydro, 5,6-dihydro-4h-pyran, dihydropyrane
PubChem CID 8080
IUPAC Name 3,4-dihydro-2H-pyran
SMILES C1CC=COC1

Specifications

Color Yellow
Boiling Point 86°C
Quantity 100 mL
UN Number 2376
Formula Weight 84.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dihydro-2H-pyran
EINECSNumber (5)-0670
TSCA Yes