missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-chromen-3-ylmethanol, 97%, Thermo Scientific™

Catalog No. CC25809CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC25809CB 250mg Amber Glass Bottle
CC25809DA 1g Amber Glass Bottle
CC25809DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC25809CB Supplier Thermo Scientific Chemicals Supplier No. CC25809CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76727-28-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD04972641
InChI Key UPDSWAQIQZFOLF-UHFFFAOYSA-N
Synonym chroman-3-ylmethanol, 3,4-dihydro-2h-1-benzopyran-3-ylmethanol, 3-hydroxymethylchroman, chroman-3-yl-methanol, chroman-3-ylmethan-1-ol, 3,4-dihydro-2h-1-benzopyran 3-methanol, 2h-1-benzopyran-3-methanol,3,4-dihydro, 2h-1-benzopyran-3-methanol, 3,4-dihydro
PubChem CID 2795460
IUPAC Name 3,4-dihydro-2H-chromen-3-ylmethanol
SMILES C1C(COC2=CC=CC=C21)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 164.2
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-chromen-3-ylmethanol