missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC13201DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC13201DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

20825-89-2
194.186
MQSSBVLREFSMDP-UHFFFAOYSA-N
2776383
C1COC2=C(C=C(C=C2)C(=O)O)OC1
C10H10O4
MFCD00276310
3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carboxylic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylic acid, 2h-1,5-benzodioxepin-7-carboxylic acid, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-carboxylic acid, acmc-1crio, 7-carboxy-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin-7-carboxylicacid, 3,4-dihydro, 2h-1,5-benzodioxepin-7-carboxylicacid,3,4-dihydro
3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid

Specifications

3, 4-Dihydro-2H-1, 5-benzodioxepine-7-carboxylic acid
96.5
97%
C10H10O4
MFCD00276310
MQSSBVLREFSMDP-UHFFFAOYSA-N
3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid
2776383
97%
20825-89-2
100.0
Amber Glass Bottle
1g
3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carboxylic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylic acid, 2h-1,5-benzodioxepin-7-carboxylic acid, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-carboxylic acid, acmc-1crio, 7-carboxy-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin-7-carboxylicacid, 3,4-dihydro, 2h-1,5-benzodioxepin-7-carboxylicacid,3,4-dihydro
C1COC2=C(C=C(C=C2)C(=O)O)OC1
194.186
194.19
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents