missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-ylmethylamine, 97%, Thermo Scientific™

Catalog No. cc13213da
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13213DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC13213DA Supplier Thermo Scientific Chemicals Supplier No. CC13213DA

Chemical Identifiers

CAS 23475-00-5
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD02682040
InChI Key MCHQABHXBPYZCT-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-1,5-benzodioxepin-7-ylmethylamine, c-3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl-methylamine, 2h-1,5-benzodioxepin-7-methanamine,3,4-dihydro, 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl methanamine, 1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl methanamine, 2h,3h,4h-benzo b 1,4-dioxepan-7-ylmethylamine, 2h,3h,4h-benzo b 1,4-dioxepin-7-ylmethylamine, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl methanamine, 3,4-dihydro-2h-1,5-benzodioxepin-7-ylmethyl amine, 3,4-dihydro-2h-1,5-benzodioxepin-7-yl-methylamine
PubChem CID 2795295
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepin-7-ylmethanamine
SMILES C1COC2=C(C=C(C=C2)CN)OC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepin-7-ylmethylamine
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.