missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde, 95%

Catalog No. p-7031186
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3216703 1 g
AAH3216706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3216703 Supplier Thermo Scientific Chemicals Supplier No. H3216703
Only null left

CAS: 67869-90-3 | C10H10O3 | 178.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67869-90-3
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD01893356
InChI Key LCSVYSVGXQQHSI-UHFFFAOYSA-N
PubChem CID 2776388
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde
SMILES C1COC2=C(C=C(C=C2)C=O)OC1

Specifications

Boiling Point 120°C to 122°C
Quantity 1 g
Synonym 2h-1,5-benzodioxepin-7-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxaldehyde, 2h-1,5-benzodioxepin-7-carboxaldehyde,3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepane-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepine-7-carboxaldehyde, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyd
Formula Weight 178.19
Percent Purity 95%
Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin-7-carboxaldehyde
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only