missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-6-ylmethylamine hydrochloride, 97%, Thermo Scientific™

$290.07 - $290.07

Chemical Identifiers

CAS 499770-91-1
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD04972615
InChI Key ZXJOKQNPRXXPJT-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-1,5-benzodioxepin-6-ylmethylamine, 3,4-dihydro-2h-benzo b 1,4 dioxepin-6-yl methanamine, 1-3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, 2h-1,5-benzodioxepin-6-methanamine,3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, vxu
PubChem CID 2794995
SMILES NCC1=C2OCCCOC2=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC00413DA
SDS View Documents
Thermo Scientific Chemicals
CC00413DA
1 g Amber Glass Bottle
Each for $290.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

499770-91-1
179.22
ZXJOKQNPRXXPJT-UHFFFAOYSA-N
2794995
C10H13NO2
MFCD04972615
3,4-dihydro-2h-1,5-benzodioxepin-6-ylmethylamine, 3,4-dihydro-2h-benzo b 1,4 dioxepin-6-yl methanamine, 1-3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, 2h-1,5-benzodioxepin-6-methanamine,3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, vxu
NCC1=C2OCCCOC2=CC=C1

Specifications

499770-91-1
100.0
Amber Glass Bottle
MFCD04972615
3,4-dihydro-2h-1,5-benzodioxepin-6-ylmethylamine, 3,4-dihydro-2h-benzo b 1,4 dioxepin-6-yl methanamine, 1-3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, 2h-1,5-benzodioxepin-6-methanamine,3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, vxu
NCC1=C2OCCCOC2=CC=C1
179.22
215.68
3, 4-Dihydro-2H-1, 5-benzodioxepin-6-ylmethylamine hydrochloride
96.5
97%
C10H13NO2
1 g
ZXJOKQNPRXXPJT-UHFFFAOYSA-N
3,4-dihydro-2H-1,5-benzodioxepin-6-ylmethanamine
2794995
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'