missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2-methoxy-4-methyl-2H-pyran (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. d205325ml
Change view
Click to view available options
Quantity:
25 mL
Catalog No. Quantity
D205325ML 25 mL
1 options

Catalog No. D205325ML

Supplier: TCI America D205325ML

Only null left

Chemical Identifiers

CAS 53608-95-0
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00143062
InChI Key JVTCUFUKBFIIBA-UHFFFAOYSA-N
PubChem CID 547114
IUPAC Name 2-methoxy-4-methyl-3,4-dihydro-2H-pyran
SMILES CC1CC(OC=C1)OC

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 mL
UN Number 1993
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dihydro-2-methoxy-4-methyl-2H-pyran (cis- and trans- mixture)