missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2(1H)-quinolinone, 98%

Catalog No. p-3759042
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398010010 1 g Glass bottle
AC398010100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398010010 Supplier Thermo Scientific Chemicals Supplier No. 398010010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 553-03-7 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 553-03-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00016722
InChI Key TZOYXRMEFDYWDQ-UHFFFAOYSA-N
Synonym 3,4-dihydro-2 1h-quinolinone, 3,4-dihydroquinolin-2 1h-one, hydrocarbostyril, 1,2,3,4-tetrahydroquinolin-2-one, 3,4-dihydrocarbostyril, 2-oxo-1,2,3,4-tetrahydroquinoline, dihydro-quinolinone, 3,4-dihydro-2-quinolinol, o-aminohydrocinnamic acid lactam, unii-2ckg6tx32f
PubChem CID 64796
IUPAC Name 3,4-dihydro-1H-quinolin-2-one
SMILES O=C1CCC2=CC=CC=C2N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 165.0°C to 167.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 4817
Formula Weight 147.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dihydro-2(1H)-quinolinone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and wat

RUO – Research Use Only