missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2-(1H)-quinolinone, 98%

Catalog No. p-7032987
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3663803 1 g
AAH3663806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3663803 Supplier Thermo Scientific Chemicals Supplier No. H3663803
Only null left

CAS: 553-03-7 | C9H9NO | 147.18 g/mol

3,4-Dihydro-2-(1H)-quinolinone may be employed as medium supplement in the culture medium of Pseudomonas ayucida during enrichment culture experiments. It is used to prepare potent bicyclic peptide deformylase inhibitors with antibacterial effects. It is also used to synthesize substituted iminopiperidines as inhibitors of human nitric oxide synthase isoforms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydro-2-(1H)-quinolinone may be employed as medium supplement in the culture medium of Pseudomonas ayucida during enrichment culture experiments. It is used to prepare potent bicyclic peptide deformylase inhibitors with antibacterial effects. It is also used to synthesize substituted iminopiperidines as inhibitors of human nitric oxide synthase isoforms.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 553-03-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00016722
InChI Key TZOYXRMEFDYWDQ-UHFFFAOYSA-N
Synonym 3,4-dihydro-2 1h-quinolinone, 3,4-dihydroquinolin-2 1h-one, hydrocarbostyril, 1,2,3,4-tetrahydroquinolin-2-one, 3,4-dihydrocarbostyril, 2-oxo-1,2,3,4-tetrahydroquinoline, dihydro-quinolinone, 3,4-dihydro-2-quinolinol, o-aminohydrocinnamic acid lactam, unii-2ckg6tx32f
PubChem CID 64796
SMILES O=C1CCC2=CC=CC=C2N1

Specifications

Melting Point 165°C to 167°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
IUPAC Name 3,4-dihydro-1H-quinolin-2-one
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 3,4-Dihydro-2-(1H)-quinolinone
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only