missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihexylthiophene 97.0+%, TCI America™

Catalog No. D42021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42021G 1 g
D42025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42021G Supplier TCI America Supplier No. D42021G
Only null left

Chemical Identifiers

CAS 122107-04-4
Molecular Formula C16H28S
Molecular Weight (g/mol) 252.46
MDL Number MFCD21363048
InChI Key YIRIIAIZQBBXHL-UHFFFAOYSA-N
PubChem CID 15360364
IUPAC Name 3,4-dihexylthiophene
SMILES CCCCCCC1=CSC=C1CCCCCC

Specifications

Color Yellow
Boiling Point 103°C
Quantity 1 g
Formula Weight 252.46
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dihexylthiophene