missing translation for 'onlineSavingsMsg'
Learn More

3,4'-Dihexyl-2,2'-bithiophene 96.0+%, TCI America™

Catalog No. p-7128002
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39281G 1 g
D39285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39281G Supplier TCI America Supplier No. D39281G
Only null left

Chemical Identifiers

CAS 135926-93-1
Molecular Formula C20H30S2
Molecular Weight (g/mol) 334.58
MDL Number MFCD15072161
InChI Key FWQMKAFKIYUBKB-UHFFFAOYSA-N
PubChem CID 11348225
IUPAC Name 3,4'-dihexyl-2,2'-bithiophene
SMILES CCCCCCC1=CSC(=C1)C1=C(CCCCCC)C=CS1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 1 g
Formula Weight 334.58
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4′-Dihexyl-2,2′-bithiophene
TSCA No
Recommended Storage Refrigerator