missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7127573
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29411G 1 g
D29415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29411G Supplier TCI America Supplier No. D29411G
Only null left

Chemical Identifiers

CAS 145758-05-0
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00153120
InChI Key FSGLUBQENACWCC-UHFFFAOYSA-N
Synonym 3,4-difluorobenzene-1-sulfonyl chloride, 3,4-difluorobenzenesulphonyl chloride, 3,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-difluoro, 3,4-difluorophenyl chlorosulfone, 3,4-difluorobenzenesulfonylchloride, 3,4-difluorobenzene sulfonyl chloride, 3,4-difluorobenzenesulphonylchloride, pubchem1958, acmc-1byq8
PubChem CID 2736913
IUPAC Name 3,4-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)F)F

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 212.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorobenzenesulfonyl Chloride